
4-(Bromomethyl)-2-fluorobenzenesulphonyl chloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1820684-91-0
:
Formula:C₇H₅BrClFO₂S
MDL Number:MFCD28096606
:287.534 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
Frequently Bought Together