3-Difluoromethyl-5-phenylpyrazole
Catalog # APOSPC4856-10G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:194.181 g/mol
Storage Temperature:Ambient
CAS Number:122980-86-3
Formula:C₁₀H₈F₂N₂
MDL Number:MFCD04039285