
Difluoromethyl 2,2,3,4,4,4-hexafluorobutyl ether 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:69948-46-5
:
Formula:C₅H₄F₈O
MDL Number:MFCD00236123
:232.072 g/mol
Boiling Pt:72…73 °C
Density:1,532 g/cm³ (25 °C)
:GHS07
Storage Temperature:Temperatura otoczenia
Frequently Bought Together