Order Entry
Poland
ContactUsLinkComponent
1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450369-25G
Supplier:  Apollo Scientific
CAS Number:  
1H,1H,2H,2H-Perfluorohexyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450369-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :590.153 g/mol
Storage Temperature:Ambient
CAS Number:1980085-36-6
Formula:C₁₃H₆F₂₀O₃
MDL Number:MFCD28334362