2,2,3,4,4,4-Hexafluorobutyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450352-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:508.128 g/mol
Storage Temperature:Ambient
CAS Number:1980086-55-2
Formula:C₁₁H₅F₁₇O₃
MDL Number:MFCD28334345