3-Fluoropropyl 1H,1H,2H,2H-perfluorohexyl carbonate
Catalog # APOSPC450349-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:368.169 g/mol
Storage Temperature:Ambient
CAS Number:1980085-32-2
Formula:C₁₀H₁₀F₁₀O₃
MDL Number:MFCD28334342