3-Fluoropropyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450346-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:404.15 g/mol
Storage Temperature:Ambient
CAS Number:1980085-56-0
Formula:C₁₀H₈F₁₂O₃
MDL Number:MFCD28334339