2,2,3,3-Tetrafluoropropyl 1H,1H-perfluorohexyl carbonate
Catalog # APOSPC450340-25G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:458.121 g/mol
Storage Temperature:Ambient
CAS Number:1980040-07-0
Formula:C₁₀H₅F₁₅O₃
MDL Number:MFCD28334333