

1H,1H-Perfluoroheptyl methacrylate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:48076-44-4
:
Formula:C₁₁H₇F₁₃O₂
MDL Number:MFCD16621324
:418.151 g/mol
UN:1993
ADR:3,III
:GHS02
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together