1H,1H-Heptafluorobutyl butyrate
Catalog # APOSPC450210-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:270.145 g/mol
Storage Temperature:Ambient
CAS Number:2926-44-5
Formula:C₈H₉F₇O₂
MDL Number:MFCD22123991