
4-(4,6-Difluoropyrimidin-2-yl)morpholine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:189003-02-9
:
Formula:C₈H₉F₂N₃O
MDL Number:MFCD00457690
:201.173 g/mol
Melting Pt:128…129 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together