
Hexakis(1H,1H-perfluorohexyloxy)phosphazene
Catalog # APOSPC450173-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1929.313 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1365808-40-7
Formula:C₃₆H₁₂F₆₆N₃O₆P₃
MDL Number:MFCD22124076