
Hexakis(1H,1H-perfluorobutoxy)phosphazene
Catalog # APOSPC450169-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:1329.223 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:470-73-5
Formula:C₂₄H₁₂F₄₂N₃O₆P₃
MDL Number:MFCD22124072