Order Entry
Poland
ContactUsLinkComponent
 
Flammable
Irritant
Bis(2,2,3,3-tetrafluoropropyl)ether
Catalog # APOSPC450069-250MG
Supplier:  Apollo Scientific
CAS Number:  
Flammable
Irritant
Bis(2,2,3,3-tetrafluoropropyl)ether
Catalog # APOSPC450069-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :246.098 g/mol
Storage Temperature:Ambient
UN:1993
ADR:3,III
 :GHS02
 :GHS07
CAS Number:82914-35-0
Formula:C₆H₆F₈O
MDL Number:MFCD22123965