Order Entry
Poland
ContactUsLinkComponent
3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile
3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile
Catalog # APOSPC450013-250MG
Supplier:  Apollo Scientific
CAS Number:  
3-(2-Iodoethyl)-4-(1H,1H-perfluoroheptyl)pyrrolidine-1-carbonitrile
Catalog # APOSPC450013-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :582.142 g/mol
Storage Temperature:Ambient
CAS Number:1365808-06-5
Formula:C₁₄H₁₂F₁₃IN₂
MDL Number:MFCD22124040