Order Entry
Poland
ContactUsLinkComponent
3,3-Difluoro-6-methoxyindolin-2-one 95%
3,3-Difluoro-6-methoxyindolin-2-one 95%
Catalog # APOSPC448200-250MG
Supplier:  Apollo Scientific
CAS Number:  
3,3-Difluoro-6-methoxyindolin-2-one 95%
Catalog # APOSPC448200-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :199.154 g/mol
Storage Temperature:Ambient
CAS Number:1393560-43-4
Formula:C₉H₇F₂NO₂
MDL Number:MFCD22547892