2-(4-Fluorophenyl)-3-nitronaphthalene
Catalog # APOSPC440012-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:267.255 g/mol
Storage Temperature:Ambient
CAS Number:1450995-50-2
Formula:C₁₆H₁₀FNO₂
MDL Number:MFCD28154811