Order Entry
Poland
ContactUsLinkComponent
(S)-3,3'-Bis(9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl-N-triflyl phosphoramide
(S)-3,3'-Bis(9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl-N-triflyl phosphoramide
Catalog # APOSPC440011-100MG
Supplier:  Apollo Scientific
CAS Number:  
(S)-3,3'-Bis(9-anthracenyl)-1,1'-binaphthyl-2,2'-diyl-N-triflyl phosphoramide
Catalog # APOSPC440011-100MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :831.792 g/mol
Melting Pt:290…300 °C
Storage Temperature:Ambient
Formula:C₄₉H₂₉F₃NO₅PS
MDL Number:MFCD28139337