Order Entry
Poland
ContactUsLinkComponent
(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
Catalog # APOSPC440002-250MG
Supplier:  Apollo Scientific
CAS Number:  
(R)-3,3'-Bis(3,5-difluorophenyl)-[1,1'-binapthalene]-2,2'-diol
Catalog # APOSPC440002-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :510.478 g/mol
Storage Temperature:Ambient
CAS Number:1706439-53-3
Formula:C₃₂H₁₈F₄O₂
MDL Number:MFCD17019237