Order Entry
Poland
ContactUsLinkComponent
2-(2-(4-(Trifluoromethyl)-2-oxopyrimidin-1(2H)-yl)ethyl)isoindoline-1,3-dione
2-(2-(4-(Trifluoromethyl)-2-oxopyrimidin-1(2H)-yl)ethyl)isoindoline-1,3-dione
Catalog # APOSPC430604-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(2-(4-(Trifluoromethyl)-2-oxopyrimidin-1(2H)-yl)ethyl)isoindoline-1,3-dione
Catalog # APOSPC430604-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :337.253 g/mol
Storage Temperature:Ambient
CAS Number:1206970-29-7
Formula:C₁₅H₁₀F₃N₃O₃
MDL Number:MFCD14585272