6-Chloro-5-(trifluoromethyl)pyridin-2-amine 98%
Catalog # APOSPC421157-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:196.558 g/mol
Storage Temperature:Ambient
CAS Number:79456-28-3
Formula:C₆H₄ClF₃N₂
MDL Number:MFCD11848573