
2-(3-Fluorophenyl)-4'-fluoroacetophenone
Catalog # APOSPC421012-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:232.225 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:370874-66-1
Formula:C₁₄H₁₀F₂O
MDL Number:MFCD11210358