Order Entry
Poland
ContactUsLinkComponent
3-(Difluoromethyl)-3-methylazetidine hydrochloride
3-(Difluoromethyl)-3-methylazetidine hydrochloride
Catalog # APOSPC411265-1G
Supplier:  Apollo Scientific
CAS Number:  
3-(Difluoromethyl)-3-methylazetidine hydrochloride
Catalog # APOSPC411265-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :157.589 g/mol
Storage Temperature:Ambient
CAS Number:2227206-51-9
Formula:C₅H₉F₂N·ClH
MDL Number:MFCD31696021