Order Entry
Poland
ContactUsLinkComponent
1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Catalog # APOSPC410466-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(1,3-Dimethyl-1H-pyrazol-4-yl)-N-(4-fluorobenzyl)methanamine
Catalog # APOSPC410466-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :233.285 g/mol
Storage Temperature:Ambient
CAS Number:1006464-99-8
Formula:C₁₃H₁₆FN₃
MDL Number:MFCD04968199