1-(Difluoromethyl)-3-nitro-1H-pyrazole
Catalog # APOSPC410307-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:163.082 g/mol
Storage Temperature:Ambient
CAS Number:1002034-26-5
Formula:C₄H₃F₂N₃O₂
MDL Number:MFCD04967410