Order Entry
Poland
ContactUsLinkComponent
1-(2,2-Difluoroethyl)piperazine dihydrochloride
1-(2,2-Difluoroethyl)piperazine dihydrochloride
Catalog # APOSPC410091-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(2,2-Difluoroethyl)piperazine dihydrochloride
Catalog # APOSPC410091-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.092 g/mol
Storage Temperature:Ambient
CAS Number:1225276-98-1
Formula:C₆H₁₂F₂N₂·2ClH
MDL Number:MFCD25371148