(S)-1-(3-Chloro-5-trifluoromethyl-pyridin-2-yl)-pyrrolidin-3-ol
Catalog # APOSPC408390-500MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:266.647 g/mol
Storage Temperature:Ambient
CAS Number:1311776-29-0
Formula:C₁₀H₁₀ClF₃N₂O
MDL Number:MFCD22631734