Order Entry
Poland
ContactUsLinkComponent
3,3-Bis(trifluoromethyl)-2-fluoro-2-(pentafluoroethyl)oxirane
Irritant
3,3-Bis(trifluoromethyl)-2-fluoro-2-(pentafluoroethyl)oxirane
Catalog # APOSPC408139-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3,3-Bis(trifluoromethyl)-2-fluoro-2-(pentafluoroethyl)oxirane
Catalog # APOSPC408139-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :316.044 g/mol
Boiling Pt:53 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:788-67-0
Formula:C₆F₁₂O
MDL Number:MFCD00155785