
3-Fluoro-5-(piperidin-3-yl)pyridine dihydrochloride 97%
Catalog # APOSPC402110-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:253.144 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:1260864-37-6
Formula:C₁₀H₁₃FN₂·2ClH
MDL Number:MFCD30099272