
(S)-2-(3,5-Difluorophenyl)piperidine hydrochloride 97%
Catalog # APOSPC402107-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:233.685 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:1212946-48-9
Formula:C₁₁H₁₃F₂N·ClH
MDL Number:MFCD26583092