
Methyl 6-(trifluoromethyl)-1H-indazole-3-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:877773-17-6
:
Formula:C₁₀H₇F₃N₂O₂
MDL Number:MFCD09878903
:244.17 g/mol
Melting Pt:240…242 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together