Order Entry
Poland
ContactUsLinkComponent
1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
Catalog # APOSPC400569-1G
Supplier:  Apollo Scientific
CAS Number:  
1-[(1,1-Dimethylethyl)dimethylsilyl]-5-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-Indole
Catalog # APOSPC400569-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :375.361 g/mol
Storage Temperature:Ambient
CAS Number:1148004-02-7
Formula:C₂₀H₃₁BFNO₂Si
MDL Number:MFCD22690780