

2-(2,2-Dibromovinyl)-1-chloro-3-fluorobenzene
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1468662-00-1
:
Formula:C₈H₄Br₂ClF
MDL Number:MFCD31561228
:314.377 g/mol
UN:2810
ADR:6.1,III
:GHS05
:GHS06
Storage Temperature:Ambient
Frequently Bought Together