Order Entry
Poland
ContactUsLinkComponent
1-(4-Fluorophenyl)cyclopropanecarbonitrile
Irritant
1-(4-Fluorophenyl)cyclopropanecarbonitrile
Catalog # APOSPC32217-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(4-Fluorophenyl)cyclopropanecarbonitrile
Catalog # APOSPC32217-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :161.176 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:97009-67-1
Formula:C₁₀H₈FN
MDL Number:MFCD01314307