Order Entry
Poland
ContactUsLinkComponent
 
5-Bromo-2,3-difluorophenylacetonitrile 97%
Catalog # APOSPC303425-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Bromo-2,3-difluorophenylacetonitrile 97%
Catalog # APOSPC303425-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.025 g/mol
Storage Temperature:Ambient
CAS Number:1780864-29-0
Formula:C₈H₄BrF₂N
MDL Number:MFCD28738339