3-Fluoro-4-methyl-5-(trifluoromethyl)benzenesulphonamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2149597-25-9
:
Formula:C₈H₇F₄NO₂S
MDL Number:MFCD30502874
:257.205 g/mol
Storage Temperature:Ambient
Frequently Bought Together