2,3-Difluoro-5-methoxy-4-methylphenylacetonitrile 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706461-19-9
:
Formula:C₁₀H₉F₂NO
MDL Number:MFCD27986852
:197.181 g/mol
Storage Temperature:Ambient
UN:3439
ADR:6.1,III
Frequently Bought Together