Order Entry
Poland
ContactUsLinkComponent
3-Ethoxy-2,6-difluorobenzenesulphonyl chloride
3-Ethoxy-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303250-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Ethoxy-2,6-difluorobenzenesulphonyl chloride
Catalog # APOSPC303250-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :256.654 g/mol
Storage Temperature:Ambient
CAS Number:1552934-62-9
Formula:C₈H₇ClF₂O₃S
MDL Number:MFCD24284208