2-Methoxy-5-(trifluoromethoxy)benzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1010422-55-5
:
Formula:C₈H₆ClF₃O₄S
MDL Number:MFCD13176696
:290.644 g/mol
Storage Temperature:Ambient
Frequently Bought Together