2,3-Dichloro-6-fluorobenzenesulphonyl chloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1706446-66-3
:
Formula:C₆H₂Cl₃FO₂S
MDL Number:MFCD28054218
:263.501 g/mol
Storage Temperature:Temperatura otoczenia
UN:1760
ADR:8,III
Frequently Bought Together