Order Entry
Poland
ContactUsLinkComponent
 
3,5-Difluoro-2-methoxybenzamide 97%
Catalog # APOSPC302939-5G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Difluoro-2-methoxybenzamide 97%
Catalog # APOSPC302939-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :187.143 g/mol
Storage Temperature:Ambient
CAS Number:874804-09-8
Formula:C₈H₇F₂NO₂
MDL Number:MFCD06409086