Order Entry
Poland
ContactUsLinkComponent
4-Ethoxy-3,5-difluorocinnamic acid 97%
4-Ethoxy-3,5-difluorocinnamic acid 97%
Catalog # APOSPC302211-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Ethoxy-3,5-difluorocinnamic acid 97%
Catalog # APOSPC302211-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :228.192 g/mol
Storage Temperature:Ambient
CAS Number:1017779-43-9
Formula:C₁₁H₁₀F₂O₃
MDL Number:MFCD09258693