Order Entry
Poland
ContactUsLinkComponent
3-Chloro-5-(trifluoromethoxy)cinnamic acid 97%
3-Chloro-5-(trifluoromethoxy)cinnamic acid 97%
Catalog # APOSPC302049-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Chloro-5-(trifluoromethoxy)cinnamic acid 97%
Catalog # APOSPC302049-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :266.601 g/mol
Storage Temperature:Ambient
CAS Number:1588505-18-3
Formula:C₁₀H₆ClF₃O₃
MDL Number:MFCD06660339