2,4-Dichloro-N-[2-methyl-3-(trifluoromethyl)phenyl]benzenesulphonamide
Catalog # APOSPC300928-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:384.201 g/mol
Storage Temperature:Ambient
Formula:C₁₄H₁₀Cl₂F₃NO₂S
MDL Number:MFCD30481168