4-[4-Amino-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Catalog # APOSPC300840-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:332.298 g/mol
Storage Temperature:Ambient
CAS Number:1858252-06-8
Formula:C₁₃H₁₁F₃N₂O₃S
MDL Number:MFCD28347996