4-[4-Nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Catalog # APOSPC300839-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:362.281 g/mol
Storage Temperature:Ambient
CAS Number:1858251-46-3
Formula:C₁₃H₉F₃N₂O₅S
MDL Number:MFCD28347994