N,N-Dimethyl-4-[4-nitro-2-(trifluoromethyl)phenoxy]benzenesulphonamide
Catalog # APOSPC300837-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:390.334 g/mol
Storage Temperature:Ambient
CAS Number:1858250-85-7
Formula:C₁₅H₁₃F₃N₂O₅S
MDL Number:MFCD28347993