4-{2-[5-(Trifluoromethyl)-1H-pyrazol-1-yl]-1,3-thiazol-4-yl}aniline
Catalog # APOSPC300667-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:310.298 g/mol
Storage Temperature:Ambient
Formula:C₁₃H₉F₃N₄S
MDL Number:MFCD12406836