Order Entry
Poland
ContactUsLinkComponent
1-(Bromomethyl)-2-(2-fluorophenoxy)benzene | 1-(Bromomethyl)-2-(2-fluorophenoxy)benzene
1-(Bromomethyl)-2-(2-fluorophenoxy)benzene
Catalog # APOSPC300608-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(Bromomethyl)-2-(2-fluorophenoxy)benzene
Catalog # APOSPC300608-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :281.12 g/mol
Storage Temperature:Ambient
CAS Number:1355171-21-9
Formula:C₁₃H₁₀BrFO
MDL Number:MFCD15146804