Order Entry
Poland
ContactUsLinkComponent
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC300601-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N-Dimethyl-4-{[4-(trifluoromethyl)pyrimidin-2-yl]oxy}benzenesulphonamide
Catalog # APOSPC300601-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :347.313 g/mol
Storage Temperature:Ambient
CAS Number:1227954-78-0
Formula:C₁₃H₁₂F₃N₃O₃S
MDL Number:MFCD11109533